MilliporeSigma
  • RDL A301S alone does not confer high levels of resistance to cyclodiene organochlorine or phenyl pyrazole insecticides in Plutella xylostella.
RDL A301S alone does not confer high levels of resistance to cyclodiene organochlorine or phenyl pyrazole insecticides in Plutella xylostella.
Pesticide biochemistry and physiology (2019-08-06)
M Guest, J A Goodchild, J A Bristow, A J Flemming
ABSTRACT
Mutations in the GABA-gated chloride channel are associated with resistance to cyclodiene organochlorine and phenyl pyrazole insecticides. The best characterised of these is A301S, which was initially identified in a Dieldrin resistant strain of Drosophila melanogaster. The orthologous mutation has been found in a variety of different crop pests including the diamond back moth Plutella xylostella. However, the contribution of this mutation to resistance in this species remains unclear. We have used the CRISPR/Cas9 system in order to edit Plutella xylostella PxGABARalpha1 to Serine at the 301 orthologous position (282 in PxGABARalpha1) in an insecticide sensitive strain isolated from Vero Beach (VB) USA. In this edited line, no high level of resistance is conferred to Dieldrin, Endosulfan or Fipronil, rather only a subtle shift in sensitivity which could not confer commercially important resistance. We conclude that the high level of commercial resistance to cyclodiene organochlorine and phenyl pyrazole insecticides observed in some field isolates of Plutella xylostella cannot arise from A282S in PxGABARalpha1 alone.