MilliporeSigma
  • Berberine chloride mediates its antileishmanial activity by inhibiting Leishmania mitochondria.
Berberine chloride mediates its antileishmanial activity by inhibiting Leishmania mitochondria.
Parasitology research (2018-11-25)
Sritama De Sarkar, Deblina Sarkar, Avijit Sarkar, Aishwarya Dighal, Katrin Staniek, Lars Gille, Mitali Chatterjee
ABSTRACT
Berberine chloride, a plant-derived isoquinoline alkaloid, has been demonstrated to have leishmanicidal activity, which is mediated by generation of a redox imbalance and depolarization of the mitochondrial membrane, resulting in a caspase-independent apoptotic-like cell death. However, its impact on mitochondrial function remains to be delineated and is the focus of this study. In UR6 promastigotes, berberine chloride demonstrated a dose-dependent increase in generation of reactive oxygen species and mitochondrial superoxide, depolarization of the mitochondrial membrane potential, a dose-dependent inhibition of mitochondrial complexes I-III and II-III, along with a substantial depletion of ATP, collectively suggesting inhibition of parasite mitochondria. Accordingly, the oxidative stress induced by berberine chloride resulting in an apoptotic-like cell death in Leishmania can be exploited as a potent chemotherapeutic strategy, mitochondria being a prime contributor.