MilliporeSigma
  • Synthesis and in vitro urease inhibitory activity of benzohydrazide derivatives, in silico and kinetic studies.
Synthesis and in vitro urease inhibitory activity of benzohydrazide derivatives, in silico and kinetic studies.
Bioorganic chemistry (2018-10-16)
Azhar Abbas, Basharat Ali, Kanwal, Khalid Mohammed Khan, Jamshed Iqbal, Shafiq Ur Rahman, Sumera Zaib, Shahnaz Perveen
ABSTRACT
Benzohydrazide derivatives 1-43 were synthesized via "one-pot" reaction and structural characterization of these synthetic derivatives was carried out by different spectroscopic techniques such as 1H NMR and EI-MS. The synthetic molecules were evaluated for their in vitro urease inhibitory activity. All synthetic derivatives showed good inhibitory activities in the range of (IC50 = 0.87 ± 0.31-19.0 ± 0.25 µM) as compared to the standard thiourea (IC50 = 21.25 ± 0.15 µM), except seven compounds 17, 18, 23, 24, 29, 30, and 41 which were found to be inactive. The most active compound of the series was compound 36 (IC50 = 0.87 ± 0.31 μM) having two chloro groups at meta positions of ring A and methoxy group at para position of ring B. The structure-activity relationship (SAR) of the active compounds was established on the basis of different substituents and their positions in the molecules. Kinetic studies of the active compounds revealed that compounds can inhibit enzyme via competitive and noncompetitive modes. In silico study was also performed to understand the binding interactions of the molecules (ligand) with the active site of enzyme.