MilliporeSigma
  • siRNA against TSG101 reduces proliferation and induces G0/G1 arrest in renal cell carcinoma - involvement of c-myc, cyclin E1, and CDK2.
siRNA against TSG101 reduces proliferation and induces G0/G1 arrest in renal cell carcinoma - involvement of c-myc, cyclin E1, and CDK2.
Cellular & molecular biology letters (2019-01-25)
Chen Xu, Junhua Zheng
ABSTRACT
The tumor susceptibility gene 101 (TSG101) is closely associated with various tumor types, but its role in the pathogenesis of renal cell carcinoma (RCC) is still unknown. This study used RNA interference to silence the expression of TSG101 in RCC cell lines and explore the role of TSG101 in RCC. Immunohistochemistry and western blot were performed to detect the expression of TSG101 in 15 paired renal tumor samples. A small interfering RNA (siRNA) targeting TSG101 was transfected into A498 and 786-O cell lines. The Cell Counting Kit-8 (CCK-8) assay and colony formation assay were used to observe the changes in cell proliferation after transfection. Flow cytometry was used to detect the effect on the cell cycle. Western blot was conducted to study the changes of related functional proteins. The expression of TSG101 was higher in RCC tissues than in adjacent normal tissues. The CCK-8 assay showed that the proliferation and colony formation of the A498 and 786-O cell lines were attenuated after suppression of TSG101. Flow cytometry showed that silencing of TSG101 induced G0/G1 arrest. The western blot results revealed that the levels of cell cycle-related proteins (c-myc, cyclin E1 and cyclin-dependent kinase 2 (CDK2)) were markedly decreased in the siRNA groups. TSG101 promotes proliferation of RCC cells. This positive effect on tumor growth involves activation of c-myc and cyclin E1/CDK2 and their effect on cell cycle distribution.