MilliporeSigma
  • Concentration of furfuryl alcohol in fluid milk, dried dairy ingredients, and cultured dairy products.
Concentration of furfuryl alcohol in fluid milk, dried dairy ingredients, and cultured dairy products.
Journal of dairy science (2019-03-05)
B M Wherry, Y Jo, M A Drake
ABSTRACT
Maillard reactions occur in dairy products during heat treatment. Furfuryl alcohol (FA) may be found in dairy products as a result of Maillard reactions. The recent posting in California Proposition 65 indicates that FA may be carcinogenic, and for this reason it is crucial to accurately measure FA concentrations in dairy products. The objective of this study was to identify an extraction and quantitation method for FA from dairy products and to determine FA concentrations in milk, dairy powders, and cultured dairy products. Solvent-assisted flavor extraction, solid-phase microextraction, stir bar sorptive extraction with gas chromatography-mass spectrometry and triple quadrupole mass spectrometry were compared for recovery of FA. Internal standards for the quantitation of FA (2-methyl-3-heptanone, furfuryl-d5 alcohol, 2,5-dimethylphenol, 5-methyl-2-furfuryl alcohol, and 5-methyl furfural) were also compared. Subsequently, fluid milk [high temperature, short time (HTST) and ultrapasteurized], whey protein isolates (3 mo-4 yr), whey protein concentrates (3 mo-4 yr), whole milk powders (1 yr), high and low heat skim milk powders (SMP; 0-8 yr), milk protein isolates (3 mo-3 yr), milk protein concentrates (3 mo-3 yr), Cheddar cheese (mild, medium, sharp, and extra sharp), mozzarella cheese (whole and part skim), cottage cheese (nonfat, low fat, and full fat), sour cream (nonfat, low fat, and full fat), traditional yogurt (nonfat, low fat, and full fat), and Greek-style yogurt (nonfat; n = 139 products total) were evaluated. Furfuryl alcohol was extracted from products by headspace solid-phase microextraction followed by gas chromatography-triple quadrupole mass spectrometry using a ZB-5ms column (30 m × 0.25 mm × 0.25 µm; Phenomenex Inc., Torrance, CA). Furfuryl-d5 alcohol was used as an internal standard. Each food was extracted in triplicate. Ultrapasteurized milks had higher levels of FA than HTST milks (122.3 vs. 7.350 µg/kg). Furfuryl alcohol concentrations ranged from 0.634 to 26.55 µg/kg in whey protein isolates, 2.251 to 56.19 µg/kg in whey protein concentrates, 11.99 to 121.9 µg/kg in milk protein isolates, and 8.312 to 49.71 µg/kg in milk protein concentrates, and concentrations increased with powder storage. High heat SMP had higher concentrations of FA than low heat SMP (11.8 vs. 1.36 µg/kg) and concentrations increased with storage time. Concentrations of FA in Cheddar and mozzarella cheese ranged from 2.361 to 110.5 µg/kg and were higher than FA concentrations in cottage cheese or sour cream (0.049-1.017 µg/kg). These results suggest that FA is present at higher levels in dairy products that have been subjected to higher temperatures or have been stored longer. Sour cream and cottage cheese had lower levels of FA. Compared with other studies on food products with reported levels of FA, such as coffee (200-400 µg/g), dairy products have very low levels of FA.