MilliporeSigma
  • Axenic Biofilm Formation and Aggregation by Synechocystis sp. Strain PCC 6803 Are Induced by Changes in Nutrient Concentration and Require Cell Surface Structures.
Axenic Biofilm Formation and Aggregation by Synechocystis sp. Strain PCC 6803 Are Induced by Changes in Nutrient Concentration and Require Cell Surface Structures.
Applied and environmental microbiology (2019-02-03)
Rey Allen, Bruce E Rittmann, Roy Curtiss
ABSTRACT
Phototrophic biofilms are key to nutrient cycling in natural environments and bioremediation technologies, but few studies describe biofilm formation by pure (axenic) cultures of a phototrophic microbe. The cyanobacterium Synechocystis sp. strain PCC 6803 (here Synechocystis) is a model microorganism for the study of oxygenic photosynthesis and biofuel production. We report here that wild-type (WT) Synechocystis caused extensive biofilm formation in a 2,000-liter outdoor nonaxenic photobioreactor under conditions attributed to nutrient limitation. We developed a biofilm assay and found that axenic Synechocystis forms biofilms of cells and extracellular material but only when cells are induced by an environmental signal, such as a reduction in the concentration of growth medium BG11. Mutants lacking cell surface structures, namely type IV pili and the S-layer, do not form biofilms. To further characterize the molecular mechanisms of cell-cell binding by Synechocystis, we also developed a rapid (8-h) axenic aggregation assay. Mutants lacking type IV pili were unable to aggregate, but mutants lacking a homolog to Wza, a protein required for type 1 exopolysaccharide export in Escherichia coli, had a superbinding phenotype. In WT cultures, 1.2× BG11 medium induced aggregation to the same degree as 0.8× BG11 medium. Overall, our data support that Wza-dependent exopolysaccharide is essential to maintain stable, uniform suspensions of WT Synechocystis cells in unmodified growth medium and that this mechanism is counteracted in a pilus-dependent manner under altered BG11 concentrations.IMPORTANCE Microbes can exist as suspensions of individual cells in liquids and also commonly form multicellular communities attached to surfaces. Surface-attached communities, called biofilms, can confer antibiotic resistance to pathogenic bacteria during infections and establish food webs for global nutrient cycling in the environment. Phototrophic biofilm formation is one of the earliest phenotypes visible in the fossil record, dating back over 3 billion years. Despite the importance and ubiquity of phototrophic biofilms, most of what we know about the molecular mechanisms, genetic regulation, and environmental signals of biofilm formation comes from studies of heterotrophic bacteria. We aim to help bridge this knowledge gap by developing new assays for Synechocystis, a phototrophic cyanobacterium used to study oxygenic photosynthesis and biofuel production. With the aid of these new assays, we contribute to the development of Synechocystis as a model organism for the study of axenic phototrophic biofilm formation.