MilliporeSigma
  • A T-cell receptor escape channel allows broad T-cell response to CD1b and membrane phospholipids.
A T-cell receptor escape channel allows broad T-cell response to CD1b and membrane phospholipids.
Nature communications (2019-01-06)
Adam Shahine, Peter Reinink, Josephine F Reijneveld, Stephanie Gras, Mira Holzheimer, Tan-Yun Cheng, Adriaan J Minnaard, John D Altman, Steffi Lenz, Jacques Prandi, Joanna Kubler-Kielb, D Branch Moody, Jamie Rossjohn, Ildiko Van Rhijn
ABSTRACT
CD1 proteins are expressed on dendritic cells, where they display lipid antigens to T-cell receptors (TCRs). Here we describe T-cell autoreactivity towards ubiquitous human membrane phospholipids presented by CD1b. These T-cells discriminate between two major types of lipids, sphingolipids and phospholipids, but were broadly cross-reactive towards diverse phospholipids including phosphatidylcholine, phosphatidylinositol and phosphatidylethanolamine. The crystal structure of a representative TCR bound to CD1b-phosphatidylcholine provides a molecular mechanism for this promiscuous recognition. We observe a lateral escape channel in the TCR, which shunted phospholipid head groups sideways along the CD1b-TCR interface, without contacting the TCR. Instead the TCR recognition site involved the neck region phosphate that is common to all major self-phospholipids but absent in sphingolipids. Whereas prior studies have focused on foreign lipids or rare self-lipids, we define a new molecular mechanism of promiscuous recognition of common self-phospholipids including those that are known targets in human autoimmune disease.
Product Description
Avanti
18:0-18:1 PG, Avanti Research - A Croda Brand