MilliporeSigma
  • Regulation of miRNAs by Snail during epithelial-to-mesenchymal transition in HT29 colon cancer cells.
Regulation of miRNAs by Snail during epithelial-to-mesenchymal transition in HT29 colon cancer cells.
Scientific reports (2019-02-17)
Patrycja Przygodzka, Izabela Papiewska-Pająk, Helena Bogusz-Koziarska, Ewelina Sochacka, Joanna Boncela, M Anna Kowalska
ABSTRACT
Epithelial-to-mesenchymal transition (EMT) in cancer cells, represents early stages of metastasis and is a promising target in colorectal cancer (CRC) therapy. There have been many attempts to identify markers and key pathways induced throughout EMT but the process is complex and depends on the cancer type and tumour microenvironment. Here we used the colon cancer cell line HT29, which stably overexpressed Snail, the key transcription factor in early EMT, as a model for colorectal adenocarcinoma cells with a pro-metastatic phenotype. We investigated miRNA expression regulation during that phenotypic switching. We found that overexpression of Snail in HT29 cells triggered significant changes in individual miRNA levels but did not change the global efficiency of miRNA processing. Snail abundance repressed the expression of miR-192 and miR-194 and increased miR-205, let-7i and SNORD13 levels. These identified changes correlated with the reported transcriptomic alterations in Snail-overexpressing HT29 cells. We also investigated how Snail affected the miRNA content of extracellular vesicles (EVs) released from HT29 cells. Our data suggest that the presence of Snail significantly alters the complex mRNA/miRNA interactions in the early steps of metastasis and also has an impact on the content of EVs released from HT29 cells.