MilliporeSigma
  • Single-dose radiotherapy disables tumor cell homologous recombination via ischemia/reperfusion injury.
Single-dose radiotherapy disables tumor cell homologous recombination via ischemia/reperfusion injury.
The Journal of clinical investigation (2018-11-28)
Sahra Bodo, Cécile Campagne, Tin Htwe Thin, Daniel S Higginson, H Alberto Vargas, Guoqiang Hua, John D Fuller, Ellen Ackerstaff, James Russell, Zhigang Zhang, Stefan Klingler, HyungJoon Cho, Matthew G Kaag, Yousef Mazaheri, Andreas Rimner, Katia Manova-Todorova, Boris Epel, Joan Zatcky, Cristian R Cleary, Shyam S Rao, Yoshiya Yamada, Michael J Zelefsky, Howard J Halpern, Jason A Koutcher, Carlos Cordon-Cardo, Carlo Greco, Adriana Haimovitz-Friedman, Evis Sala, Simon N Powell, Richard Kolesnick, Zvi Fuks
ABSTRACT
Tumor cure with conventional fractionated radiotherapy is 65%, dependent on tumor cell-autonomous gradual buildup of DNA double-strand break (DSB) misrepair. Here we report that single-dose radiotherapy (SDRT), a disruptive technique that ablates more than 90% of human cancers, operates a distinct dual-target mechanism, linking acid sphingomyelinase-mediated (ASMase-mediated) microvascular perfusion defects to DNA unrepair in tumor cells to confer tumor cell lethality. ASMase-mediated microcirculatory vasoconstriction after SDRT conferred an ischemic stress response within parenchymal tumor cells, with ROS triggering the evolutionarily conserved SUMO stress response, specifically depleting chromatin-associated free SUMO3. Whereas SUMO3, but not SUMO2, was indispensable for homology-directed repair (HDR) of DSBs, HDR loss of function after SDRT yielded DSB unrepair, chromosomal aberrations, and tumor clonogen demise. Vasoconstriction blockade with the endothelin-1 inhibitor BQ-123, or ROS scavenging after SDRT using peroxiredoxin-6 overexpression or the SOD mimetic tempol, prevented chromatin SUMO3 depletion, HDR loss of function, and SDRT tumor ablation. We also provide evidence of mouse-to-human translation of this biology in a randomized clinical trial, showing that 24 Gy SDRT, but not 3×9 Gy fractionation, coupled early tumor ischemia/reperfusion to human cancer ablation. The SDRT biology provides opportunities for mechanism-based selective tumor radiosensitization via accessing of SDRT/ASMase signaling, as current studies indicate that this pathway is tractable to pharmacologic intervention.
Sigma-Aldrich
Anti-MDC1 Antibody, clone P2B11, clone P2B11, from mouse
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse