MilliporeSigma
  • Synaptic Depotentiation and mGluR5 Activity in the Nucleus Accumbens Drive Cocaine-Primed Reinstatement of Place Preference.
Synaptic Depotentiation and mGluR5 Activity in the Nucleus Accumbens Drive Cocaine-Primed Reinstatement of Place Preference.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2019-04-06)
Michael A Benneyworth, Matthew C Hearing, Anders J Asp, Aric Madayag, Anna E Ingebretson, Clare E Schmidt, Keelia A Silvis, Erin B Larson, Stephanie R Ebner, Mark J Thomas
ABSTRACT
Understanding the neurobiological processes that incite drug craving and drive relapse has the potential to help target efforts to treat addiction. The NAc serves as a critical substrate for reward and motivated behavior, in part due to alterations in excitatory synaptic strength within cortical-accumbens pathways. The present studies investigated a causal link between cocaine-induced reinstatement of conditioned place preference and rapid reductions of cocaine-dependent increases in NAc shell synaptic strength in male mice. Cocaine-conditioned place preference behavior and ex vivo whole-cell electrophysiology showed that cocaine-primed reinstatement and synaptic depotentiation were disrupted by inhibiting AMPAR internalization via intra-NAc shell infusion of a Tat-GluA23Y peptide. Furthermore, reinstatement was driven by an mGluR5-dependent reduction in AMPAR signaling. Intra-NAc shell infusion of the mGluR5 antagonist MTEP blocked cocaine-primed reinstatement and corresponding depotentiation, whereas infusion of the mGluR5 agonist CHPG itself promoted reinstatement and depotentiated synaptic strength in the NAc shell. Optogenetic examination of circuit-specific plasticity showed that inhibition of infralimbic cortical input to the NAc shell blocked cocaine-primed reinstatement, whereas low-frequency stimulation (10 Hz) of this pathway in the absence of cocaine triggered a reduction in synaptic strength akin to that observed with cocaine, and was sufficient to promote reinstatement in the absence of a cocaine challenge. These data support a model in which mGluR5-mediated reduction in GluA2-containing AMPARs at NAc shell synapses receiving input from the infralimbic cortex is a critical factor in triggering reinstatement of cocaine-primed conditioned approach behavior.SIGNIFICANCE STATEMENT These studies identified a sequence of neural events whereby reexposure to cocaine activates a signaling cascade that alters synaptic strength in the NAc shell and triggers a behavioral response driven by a drug-associated memory.