MilliporeSigma
  • The Different Outcomes of Electrochemical Copolymerisation: 3-Hexylthiophene with Indole, Carbazole or Fluorene.
The Different Outcomes of Electrochemical Copolymerisation: 3-Hexylthiophene with Indole, Carbazole or Fluorene.
Polymers (2019-04-10)
Karolina Gebka, Tomasz Jarosz, Agnieszka Stolarczyk
ABSTRACT
Electrochemical polymerisation is reported to be a method for readily producing copolymers of various conjugated molecules. We employed this method for mixtures of indole, carbazole or fluorene with 3-hexylthiophene (HT), in order to obtain their soluble copolymers. Although polymer films were obtained, infrared (IR) and Raman investigations showed that instead of the expected linear copolymers, polyindole and polycarbazole N-substituted with HT, as well as a poly(3-hexylthiophene) (PHT)/polyfluorene blend were produced instead. Boron trifluoride diethyl etherate was also used in an attempt to promote copolymerisation, but the produced deposits were found to be highly degraded.