MilliporeSigma
  • Non-coding cis-element of Period2 is essential for maintaining organismal circadian behaviour and body temperature rhythmicity.
Non-coding cis-element of Period2 is essential for maintaining organismal circadian behaviour and body temperature rhythmicity.
Nature communications (2019-06-14)
Masao Doi, Hiroyuki Shimatani, Yuta Atobe, Iori Murai, Hida Hayashi, Yukari Takahashi, Jean-Michel Fustin, Yoshiaki Yamaguchi, Hiroshi Kiyonari, Nobuya Koike, Kazuhiro Yagita, Choogon Lee, Manabu Abe, Kenji Sakimura, Hitoshi Okamura
ABSTRACT
Non-coding cis-regulatory elements are essential determinants of development, but their exact impacts on behavior and physiology in adults remain elusive. Cis-element-based transcriptional regulation is believed to be crucial for generating circadian rhythms in behavior and physiology. However, genetic evidence supporting this model is based on mutations in the protein-coding sequences of clock genes. Here, we report generation of mutant mice carrying a mutation only at the E'-box cis-element in the promoter region of the core clock gene Per2. The Per2 E'-box mutation abolishes sustainable molecular clock oscillations and renders circadian locomotor activity and body temperature rhythms unstable. Without the E'-box, Per2 messenger RNA and protein expression remain at mid-to-high levels. Our work delineates the Per2 E'-box as a critical nodal element for keeping sustainable cell-autonomous circadian oscillation and reveals the extent of the impact of the non-coding cis-element in daily maintenance of animal locomotor activity and body temperature rhythmicity.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-mPER1 (residues 6-21) Antibody, serum, from rabbit