MilliporeSigma
  • Effect of sodium chloride concentration on off-flavor removal correlated to glucosinolate degradation and red radish anthocyanin stability.
Effect of sodium chloride concentration on off-flavor removal correlated to glucosinolate degradation and red radish anthocyanin stability.
Journal of food science and technology (2019-03-25)
Wentian Chen, Eric Karangwa, Jingyang Yu, Shuqin Xia, Biao Feng, Xiaoming Zhang
ABSTRACT
Anthocyanin-rich concentrates from different red radish can be used as natural food colorants. However, the development of off-flavor during extraction has been major challenge in processing industries. This work aimed to evaluate the effect of sodium chloride (NaCl) concentration in phosphoric acidified medium pH 2.5 on removal of off-flavor from red radish anthocyanin. The effect of NaCl concentration on anthocyanin properties was also evaluated. Results showed that the total glucosinolate was highly degraded at high NaCl concentration (< 500 mM) compared with control, leading to higher off-flavor development. Additionally, the glucosinolate degradation was positively and significantly correlated to isothiocyanate, while was negatively and significantly correlated with dimethyl di-, trisulfide, cedrol, triacetin, 6-methyl-5-hepten-2-one. Moreover, total monomeric and color properties of extracted anthocyanins were degraded at high NaCl concentration (< 500 mM) compared with control. The tentative anthocyanin identification by UPLC-TQ-MS showed 12 glycosylated anthocyanins substituted at C3 and C5 in tested anthocyanin extracts. In conclusion, higher NaCl concentration (< 500 mM) could not be useful for red radish off-flavor removal and anthocyanin properties.