MilliporeSigma
Structural Insights into the Lipid A Transport Pathway in MsbA.
Structure (London, England : 1993) (2019-05-28)
Pius S Padayatti, Sung Chang Lee, Robyn L Stanfield, Po-Chao Wen, Emad Tajkhorshid, Ian A Wilson, Qinghai Zhang
ABSTRACT
MsbA is an essential ATP-binding cassette transporter in Gram-negative bacteria that transports lipid A and lipopolysaccharide from the cytoplasmic leaflet to the periplasmic leaflet of the inner membrane. Here we report the X-ray structure of MsbA from Salmonella typhimurium at 2.8-Å resolution in an inward-facing conformation after cocrystallization with lipid A and using a stabilizing facial amphiphile. The structure displays a large amplitude opening in the transmembrane portal, which is likely required for lipid A to pass from its site of synthesis into the protein-enclosed transport pathway. Putative lipid A density is observed further inside the transmembrane cavity, consistent with a trap and flip model. Additional electron density attributed to lipid A is observed near an outer surface cleft at the periplasmic ends of the transmembrane helices. These findings provide new structural insights into the lipid A transport pathway through comparative analysis with existing MsbA structures.
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Pricing and availability is not currently available.