MilliporeSigma
  • Electrocatalysis of a Europium-Dependent Bacterial Methanol Dehydrogenase with Its Physiological Electron-Acceptor Cytochrome cGJ.
Electrocatalysis of a Europium-Dependent Bacterial Methanol Dehydrogenase with Its Physiological Electron-Acceptor Cytochrome cGJ.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-03-26)
Palraj Kalimuthu, Lena J Daumann, Arjan Pol, Huub J M Op den Camp, Paul V Bernhardt
ABSTRACT
We report the first electrochemical study of a lanthanoid-dependent methanol dehydrogenase (Eu-MDH) from the acidophilic verrucomicrobial methanotroph Methylacidiphilum fumariolicum SolV with its own physiological cytochrome cGJ electron acceptor. Eu-MDH harbours a redox active 2,7,9-tricarboxypyrroloquinoline quinone (PQQ) cofactor which is non-covalently bound but coordinates trivalent lanthanoid elements including Eu3+ . Eu-MDH and the cytochrome were co-adsorbed with the biopolymer chitosan and cast onto a mercaptoundecanol (MU) monolayer modified Au working electrode. Cyclic voltammetry of cytochrome cGJ reveals a well-defined quasi-reversible FeIII/II redox couple at +255 mV vs. NHE at pH 7.5 and this response is pH independent. The reversible one-electron response of the cytochrome cGJ transforms into a sigmoidal catalytic wave in the presence of Eu-MDH and its substrates (methanol or formaldehyde). The catalytic current was pH-dependent and pH 7.3 was found to be optimal. Kinetic parameters (pH dependence, activation energy) obtained by electrochemistry show the same trends as those obtained from an artificial phenazine ethosulfate/dichlorophenol indophenol assay.