MilliporeSigma
  • Transmission Properties of FeCl3-Intercalated Graphene and WS2 Thin Films for Terahertz Time-Domain Spectroscopy Applications.
Transmission Properties of FeCl3-Intercalated Graphene and WS2 Thin Films for Terahertz Time-Domain Spectroscopy Applications.
Nanoscale research letters (2019-07-11)
Maria O Zhukova, Benjamin T Hogan, Egor N Oparin, Polina S Shaban, Yaroslav V Grachev, Evgeniya Kovalska, Kieran K Walsh, Monica F Craciun, Anna Baldycheva, Anton N Tcypkin
ABSTRACT
Time-resolved terahertz spectroscopy has become a common method both for fundamental and applied studies focused on improving the quality of human life. However, the issue of finding materials applicable in these systems is still relevant. One of the appropriate solution is 2D materials. Here, we demonstrate the transmission properties of unique graphene-based structures with iron trichloride FeCl3 dopant on glass, sapphire and Kapton polyimide film substrates that previously were not investigated in the framework of the above-described problems in near infrared and THz ranges. We also show properties of a thin tungsten disulfide WS2 film fabricated from liquid crystal solutions transferred to a polyimide and polyethylene terephthalate substrates. The introduction of impurities, the selection of structural dimensions and the use of an appropriate substrate for modified 2D layered materials allow to control the transmission of samples for both the terahertz and infrared ranges, which can be used for creation of effective modulators and components for THz spectroscopy systems.