MilliporeSigma
  • Darunavir Population Pharmacokinetic Model Based on HIV Outpatient Data.
Darunavir Population Pharmacokinetic Model Based on HIV Outpatient Data.
Therapeutic drug monitoring (2018-11-30)
Alper Daskapan, Quynh T D Tran, Dario Cattaneo, Cristina Gervasoni, Chiara Resnati, Ymkje Stienstra, Wouter F W Bierman, Jos G W Kosterink, Tjip S van der Werf, Johannes H Proost, Jan-Willem C Alffenaar, Daniel J Touw
ABSTRACT
Darunavir is a second-generation protease inhibitor and is registered for the treatment of HIV-1 infection. The aim of this study was to develop and validate a darunavir population pharmacokinetic model based on data from daily practice. Data sets were obtained from 2 hospitals: ASST Fatebenefratelli Sacco University Hospital, Italy (hospital A), and University Medical Center Groningen, the Netherlands (hospital B). A pharmacokinetic model was developed using data from the largest data set using the iterative two-stage Bayesian procedure within the MWPharm software package. External validation was conducted using data from the smaller data set with Passing-Bablok regression and Bland-Altman analyses. In total, data from 198 patients from hospital A and 170 patients from hospital B were eligible for inclusion. A 1-compartment model with first-order absorption and elimination resulted in the best model. The Passing-Bablok analysis demonstrated a linear correlation between measured concentration and predicted concentration with r = 0.97 (P < 0.05). The predicted values correlated well with the measured values as determined by a Bland-Altman analysis and were overestimated by a mean value of 0.12 mg/L (range 0.23-0.94 mg/L). A total of 98.2% of the predicted values were within the limits of agreement. A robust population pharmacokinetic model was developed, which can support therapeutic drug monitoring of darunavir in daily outpatient settings.