MilliporeSigma
  • Chemical Constituents and Antidepressant-Like Effects in Ovariectomized Mice of the Ethanol Extract of Alternanthera philoxeroides.
Chemical Constituents and Antidepressant-Like Effects in Ovariectomized Mice of the Ethanol Extract of Alternanthera philoxeroides.
Molecules (Basel, Switzerland) (2018-09-12)
Charinya Khamphukdee, Orawan Monthakantirat, Yaowared Chulikhit, Suradet Buttachon, Michael Lee, Artur M S Silva, Nazim Sekeroglu, Anake Kijjoa
ABSTRACT
The previously unreported flavone glycoside, demethyltorosaflavone B (2) and the E-propenoic acid substituted flavone, torosaflavone E (3a), were isolated together with nine previously reported metabolites, including indole-3-carbaldehyde, oleanonic acid, vanillic acid, p-hydroxybenzoic acid, altheranthin (1a), alternanthin B (1b), demethyltorosaflavone D (3b), luteolin 8-C-E-propenoic acid (4) and chrysoeriol 7-O-rhamnoside (5), from the ethanol extract of the aerial part of Althernanthera philoxeroides. The crude ethanol extract was evaluated for its in vitro estrogenic activity in MCF-7 breast cancer cell line. The crude ethanol extract was also investigated in vivo for its antidepressant-like effects on ovariectomized mice using tail suspension and forced swimming tests, while its effect on the locomotor activity was evaluated by a Y-maze test. The effect of the crude extract on the serum corticosterone level, size and volume of uterus of the ovariectomized mice were also investigated. The expression of the mouse cyclic adenosine monophosphate (cAMP) response element-binding protein (CREB), brain-derived neurotrophic factor (BDNF) and β-actin mRNAs in hippocampus and frontal cortex was also evaluated, using semiquantitative reverse transcription-polymerase chain reaction. The crude extract and the isolated compounds 1a, 1b, 3a, 3b and 5, were evaluated for their inhibitory effects on monoamine oxidases (MAOs)-A and -B.