MilliporeSigma
  • Caspase-8 Collaborates with Caspase-11 to Drive Tissue Damage and Execution of Endotoxic Shock.
Caspase-8 Collaborates with Caspase-11 to Drive Tissue Damage and Execution of Endotoxic Shock.
Immunity (2018-07-19)
Pratyusha Mandal, Yanjun Feng, John D Lyons, Scott B Berger, Shunsuke Otani, Alexandra DeLaney, Gregory K Tharp, Kristal Maner-Smith, Eileen M Burd, Michelle Schaeffer, Sandra Hoffman, Carol Capriotti, Linda Roback, Cedrick B Young, Zhe Liang, Eric A Ortlund, Nelson C DiPaolo, Steven Bosinger, John Bertin, Peter J Gough, Igor E Brodsky, Craig M Coopersmith, Dmitry M Shayakhmetov, Edward S Mocarski
ABSTRACT
The execution of shock following high dose E. coli lipopolysaccharide (LPS) or bacterial sepsis in mice required pro-apoptotic caspase-8 in addition to pro-pyroptotic caspase-11 and gasdermin D. Hematopoietic cells produced MyD88- and TRIF-dependent inflammatory cytokines sufficient to initiate shock without any contribution from caspase-8 or caspase-11. Both proteases had to be present to support tumor necrosis factor- and interferon-β-dependent tissue injury first observed in the small intestine and later in spleen and thymus. Caspase-11 enhanced the activation of caspase-8 and extrinsic cell death machinery within the lower small intestine. Neither caspase-8 nor caspase-11 was individually sufficient for shock. Both caspases collaborated to amplify inflammatory signals associated with tissue damage. Therefore, combined pyroptotic and apoptotic signaling mediated endotoxemia independently of RIPK1 kinase activity and RIPK3 function. These observations bring to light the relevance of tissue compartmentalization to disease processes in vivo where cytokines act in parallel to execute diverse cell death pathways.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Anti-GSDMD (126-138) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution