MilliporeSigma
  • Paraneoplastic cerebellar degeneration: Yo antibody alters mitochondrial calcium buffering capacity.
Paraneoplastic cerebellar degeneration: Yo antibody alters mitochondrial calcium buffering capacity.
Neuropathology and applied neurobiology (2018-04-22)
D Panja, C A Vedeler, M Schubert
ABSTRACT
Neurodegeneration is associated with dysfunction of calcium buffering capacity and thereby sustained cellular and mitochondrial calcium overload. Paraneoplastic cerebellar degeneration (PCD), characterized by progressive Purkinje neurone degeneration following paraneoplastic Yo antibody internalization and binding to cerebellar degeneration-related protein CDR2 and CDR2L, has been linked to intracellular calcium homeostasis imbalance due to calbindin D28k malfunction. Therefore, we hypothesized that Yo antibody internalization affects not only calbindin calcium binding capacity, but also calcium-sensitive mitochondrial-associated signalling, causing mitochondrial calcium overload and thereby Purkinje neurone death. Immunohistochemically, we evaluated cerebellar organotypic slice cultures of rat brains after inducing PCD through the application of Yo antibody-positive PCD patient sera or purified antibodies against CDR2 and CDR2L how pharmacologically biased mitochondrial signalling affected PCD pathology. We found that Yo antibody internalization into Purkinje neurons caused depletion of Purkinje neurone calbindin-immunoreactivity, cannabinoid 1 receptor over-activation and alterations in the actions of the mitochondria permeability transition pore (MPTP), voltage-dependent anion channels, reactive oxygen species (ROS) and Na+ /Ca2+ exchangers (NCX). The pathological mechanisms caused by Yo antibody binding to CDR2 or CDR2L differed between the two targets. Yo-CDR2 binding did not alter the mitochondrial calcium retention capacity, cyclophilin D-independent opening of MPTP or activity of NCX. These findings suggest that minimizing intracellular calcium overload toxicity either directly with cyclosporin-A or indirectly with cannabidiol or the ROS scavenger butylated hydroxytoluene promotes mitochondrial calcium homeostasis and may therefore be used as future neuroprotective therapy for PCD patients.
Sigma-Aldrich
Anti-CDR2L antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Anti-CDR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid