MilliporeSigma
  • Influence of Boron, Tungsten and Molybdenum Modifiers on Zirconia Based Pt Catalyst for Glycerol Valorization.
Influence of Boron, Tungsten and Molybdenum Modifiers on Zirconia Based Pt Catalyst for Glycerol Valorization.
Nanomaterials (Basel, Switzerland) (2019-04-17)
Manuel Checa, Vicente Montes, Jesús Hidalgo-Carrillo, Alberto Marinas, Francisco J Urbano
ABSTRACT
The influence of boron, tungsten and molybdenum modifiers on zirconia-based Pt catalyst was studied for glycerol valorization. Zirconia modified supports were prepared by impregnation of ZrO₂ with either boric, silicontungstic or phosphomolybdic acids to obtain supports with enhanced Brönsted acidic properties. The modified supports were subsequently impregnated with chloroplatinic acid to obtain Pt-based catalysts. Pt incorporation resulted in the increase in Lewis acidity of the solids, being more significant for the Pt//W/ZrO₂ catalyst. Reduced Pt catalysts were tested for the liquid-phase glycerol hydrogenolysis, observing a synergistic effect between catalyst acid sites and metal function that proved to be crucial in glycerol hydrogenolysis. The Pt//W/ZrO₂ catalyst was the most active catalyst in this reaction, being the only leading to 1,3-PDO (45% sel., 160 °C) while Pt//Mo/ZrO₂ is the best option for 1,2-PDO (49% sel., 180 °C). Reusability studies carried out for Pt//W/ZrO₂ showed that catalytic activity dropped after the first use, remaining constant for the second and subsequent ones. Selectivity to reaction products also changes during reuses. Therefore, the selectivity to 1,2 PDO increases in the first reuse in detriment to the selectivity to n-propanol whereas the selectivity to 1,3-PDO remains constant along the uses. This behavior could be associated to the lixiviation of W species and/or catalyst fouling during reaction runs.