MilliporeSigma
  • Role of GS28 in sodium nitroprusside-induced cell death in cervical carcinoma cells.
Role of GS28 in sodium nitroprusside-induced cell death in cervical carcinoma cells.
Journal of biochemical and molecular toxicology (2019-05-09)
Do Eun Rim, Hyung Jae Yoo, Jeong-Hwa Lee, Oh-Joo Kwon, Seong-Whan Jeong
ABSTRACT
Golgi S-nitro-N-acetylpenicillamine receptor complex 1 (GS28) has been implicated in Golgi vesicle transport. We examined the role of GS28 and its molecular mechanisms in sodium nitroprusside (SNP)-induced cell death using GS28 siRNA (siGS28)-transfected HeLa cells. Significant inhibition of cytotoxicity was observed in the cells treated with SNP, and photodegraded SNP showed equal cytotoxicity to SNP. Pretreatment with an ERK inhibitor or siErk1 cotransfection blocked the inhibition in cytotoxicity. Additionally, increased phosphorylation of ERK was maintained in the cells treated with SNP, and Nrf2 level was dependent on ERK phosphorylation. However, pretreatment with a pan-caspase inhibitor had no effect on cytotoxicity or procaspase-3 level. Pretreatment with an autophagy inhibitor or siATG5 cotransfection blocked the inhibition of cytotoxicity. The changes of LC3 corresponded to that in siErk1-cotransfected cells. These data suggest that GS28 has an inductive role in SNP-induced cell death via inhibition of ERK, leading to inhibition of autophagic processes in HeLa cells.