MilliporeSigma
  • New 4-phenylcoumarin derivatives as potent 3C protease inhibitors: Design, synthesis, anti-HAV effect and molecular modeling.
New 4-phenylcoumarin derivatives as potent 3C protease inhibitors: Design, synthesis, anti-HAV effect and molecular modeling.
European journal of medicinal chemistry (2019-03-08)
Asmaa F Kassem, Rasha Z Batran, Eman M H Abbas, Samia A Elseginy, Mohamed N F Shaheen, Elmahdy M Elmahdy
ABSTRACT
A new series of 4-phenylcoumarin derivatives was synthesized starting from (2-oxo-4-phenyl-2H-chromen-7-yloxy) acetic acid hydrazide 3. Evaluation of the target compounds for their antiviral activity against hepatitis A virus revealed that the ethylthiosemicarbazide derivative 7b was the most potent virucidal agent (IC50 = 3.1 μg/ml, TI = 83). The Schiff's bases 14c and 14b demonstrated the highest virustatic effects against viral adsorption and replication, respectively (14c; IC50 = 8.5 μg/ml, TI = 88 and 14b; IC50 = 10.7 μg/ml, TI = 91). Furthermore, compounds 7b, 14b and 14c were tested against HAV 3C protease and showed significant inhibition effects (Ki = 1.903, 0.104 and 0.217 μM, respectively). The remarkable inhibitory effect expressed by the three target compounds against HAV 3C protease prompted us to expand our research on HRV 3C protease, a structurally related enzyme of the same family, and interestingly, the three target compounds displayed significant inhibitory effect against HRV 3C protease (IC50 = 16.10, 4.13 and 6.30 μM, respectively). Moreover, the active compounds 7b, 14b and 14c were docked within the pocket site of HAV 3C protease (PDB code: 2HAL) illustrating a strong H-profile with the key amino acids Gly170 and Cys172 similar to the co-crystallized ligand. Furthermore, 3D-pharmacophore and quantitative structure activity relationship (QSAR) models were generated to explore the structural requirements for the observed antiviral activity.