MilliporeSigma
  • Growth-inhibition of cell lines derived from B cell lymphomas through antagonism of serotonin receptor signaling.
Growth-inhibition of cell lines derived from B cell lymphomas through antagonism of serotonin receptor signaling.
Scientific reports (2019-03-14)
Shrikant S Kolan, Tommy Lidström, Tomás Mediavilla, Andy Dernstedt, Sofie Degerman, Magnus Hultdin, Karl Björk, Daniel Marcellino, Mattias N E Forsell
ABSTRACT
A majority of lymphomas are derived from B cells and novel treatments are required to treat refractory disease. Neurotransmitters such as serotonin and dopamine influence activation of B cells and the effects of a selective serotonin 1A receptor (5HT1A) antagonist on growth of a number of B cell-derived lymphoma cell lines were investigated. We confirmed the expression of 5HT1A in human lymphoma tissue and in several well-defined experimental cell lines. We discovered that the pharmacological inhibition of 5HT1A led to the reduced proliferation of B cell-derived lymphoma cell lines together with DNA damage, ROS-independent caspase activation and apoptosis in a large fraction of cells. Residual live cells were found 'locked' in a non-proliferative state in which a selective transcriptional and translational shutdown of genes important for cell proliferation and metabolism occurred (e.g., AKT, GSK-3β, cMYC and p53). Strikingly, inhibition of 5HT1A regulated mitochondrial activity through a rapid reduction of mitochondrial membrane potential and reducing dehydrogenase activity. Collectively, our data suggest 5HT1A antagonism as a novel adjuvant to established cancer treatment regimens to further inhibit lymphoma growth.
Sigma-Aldrich
WAY-100635 maleate salt, powder