MilliporeSigma
  • Basement membrane-like structures containing NTH α1(IV) are formed around the endothelial cell network in a novel in vitro angiogenesis model.
Basement membrane-like structures containing NTH α1(IV) are formed around the endothelial cell network in a novel in vitro angiogenesis model.
American journal of physiology. Cell physiology (2019-06-13)
Yongchol Shin, Akane Moriya, Yuta Tohnishi, Takafumi Watanabe, Yasutada Imamura
ABSTRACT
Angiogenesis is a process through which new blood vessels are formed by sprouting and elongating from existing blood vessels. Several methods have been used to replicate angiogenesis in vitro, including culturing vascular endothelial cells on Matrigel and coculturing with endothelial cells and fibroblasts. However, the angiogenesis elongation process has not been completely clarified in these models. We therefore propose a new in vitro model of angiogenesis, suitable for observing vascular elongation, by seeding a spheroid cocultured from endothelial cells and fibroblasts into a culture dish. In this model, endothelial cells formed tubular networks elongated from the spheroid with a lumen structure and were connected with tight junctions. A basement membrane (BM)-like structure was observed around the tubular network, similarly to blood vessels in vivo. These results suggested that blood vessel-like structure could be reconstituted in our model. Laminin and type IV collagen, main BM components, were highly localized around the network, along with nontriple helical form of type IV collagen α1-chain [NTH α1(IV)]. In an ascorbic acid-depleted condition, laminin and NTH α1(IV) were observed around the network but not the triple-helical form of type IV collagen and the network was unstable. These results suggest that laminin and NTH α1(IV) are involved in the formation of tubular network and type IV collagen is necessary to stabilize the network.