MilliporeSigma
  • Neuronal susceptibility to beta-amyloid toxicity and ischemic injury involves histone deacetylase-2 regulation of endophilin-B1.
Neuronal susceptibility to beta-amyloid toxicity and ischemic injury involves histone deacetylase-2 regulation of endophilin-B1.
Brain pathology (Zurich, Switzerland) (2018-07-22)
David B Wang, Chizuru Kinoshita, Yoshito Kinoshita, Bryce L Sopher, Takuma Uo, Rona J Lee, Joon Kyu Kim, Sean P Murphy, C Dirk Keene, Gwenn A Garden, Richard S Morrison
ABSTRACT
Histone deacetylases (HDACs) catalyze acetyl group removal from histone proteins, leading to altered chromatin structure and gene expression. HDAC2 is highly expressed in adult brain, and HDAC2 levels are elevated in Alzheimer's disease (AD) brain. We previously reported that neuron-specific splice isoforms of Endophilin-B1 (Endo-B1) promote neuronal survival, but are reduced in human AD brain and mouse models of AD and stroke. Here, we demonstrate that HDAC2 suppresses Endo-B1 expression. HDAC2 knockdown or knockout enhances expression of Endo-B1. Conversely, HDAC2 overexpression decreases Endo-B1 expression. We also demonstrate that neurons exposed to beta-amyloid increase HDAC2 and reduce histone H3 acetylation while HDAC2 knockdown prevents Aβ induced loss of histone H3 acetylation, mitochondrial dysfunction, caspase-3 activation, and neuronal death. The protective effect of HDAC2 knockdown was abrogated by Endo-B1 shRNA and in Endo-B1-null neurons, suggesting that HDAC2-induced neurotoxicity is mediated through suppression of Endo-B1. HDAC2 overexpression also modulates neuronal expression of mitofusin2 (Mfn2) and mitochondrial fission factor (MFF), recapitulating the pattern of change observed in AD. HDAC2 knockout mice demonstrate reduced injury in the middle cerebral artery occlusion with reperfusion (MCAO/R) model of cerebral ischemia demonstrating enhanced neuronal survival, minimized loss of Endo-B1, and normalized expression of Mfn2. These findings support the hypothesis that HDAC2 represses Endo-B1, sensitizing neurons to mitochondrial dysfunction and cell death in stroke and AD.