MilliporeSigma
  • Neuroendocrine differentiation of prostate cancer leads to PSMA suppression.
Neuroendocrine differentiation of prostate cancer leads to PSMA suppression.
Endocrine-related cancer (2018-11-08)
Martin K Bakht, Iulian Derecichei, Yinan Li, Rosa-Maria Ferraiuolo, Mark Dunning, So Won Oh, Abdulkadir Hussein, Hyewon Youn, Keith F Stringer, Chang Wook Jeong, Gi Jeong Cheon, Cheol Kwak, Keon Wook Kang, Alastair D Lamb, Yuzhuo Wang, Xuesen Dong, Lisa A Porter
ABSTRACT
Prostate-specific membrane antigen (PSMA) is overexpressed in most prostate adenocarcinoma (AdPC) cells and acts as a target for molecular imaging. However, some case reports indicate that PSMA-targeted imaging could be ineffectual for delineation of neuroendocrine (NE) prostate cancer (NEPC) lesions due to the suppression of the PSMA gene (FOLH1). These same reports suggest that targeting somatostatin receptor type 2 (SSTR2) could be an alternative diagnostic target for NEPC patients. This study evaluates the correlation between expression of FOLH1, NEPC marker genes and SSTR2. We evaluated the transcript abundance for FOLH1 and SSTR2 genes as well as NE markers across 909 tumors. A significant suppression of FOLH1 in NEPC patient samples and AdPC samples with high expression of NE marker genes was observed. We also investigated protein alterations of PSMA and SSTR2 in an NE-induced cell line derived by hormone depletion and lineage plasticity by loss of p53. PSMA is suppressed following NE induction and cellular plasticity in p53-deficient NEPC model. The PSMA-suppressed cells have more colony formation ability and resistance to enzalutamide treatment. Conversely, SSTR2 was only elevated following hormone depletion. In 18 NEPC patient-derived xenograft (PDX) models we find a significant suppression of FOLH1 and amplification of SSTR2 expression. Due to the observed FOLH1-supressed signature of NEPC, this study cautions on the reliability of using PMSA as a target for molecular imaging of NEPC. The observed elevation of SSTR2 in NEPC supports the possible ability of SSTR2-targeted imaging for follow-up imaging of low PSMA patients and monitoring for NEPC development.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-FOLH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution