MilliporeSigma
  • FUT8 drives the proliferation and invasion of trophoblastic cells via IGF-1/IGF-1R signaling pathway.
FUT8 drives the proliferation and invasion of trophoblastic cells via IGF-1/IGF-1R signaling pathway.
Placenta (2019-02-05)
Ming Yu, Xinyuan Cui, Hao Wang, Jianwei Liu, Huamin Qin, Shuai Liu, Qiu Yan
ABSTRACT
Trophoblast proliferation and invasion are essential for embryo implantation and placentation. Protein glycosylation is one of the most common and vital post-translational modifications, regulates protein physical and biochemical properties. FUT8 is the only known fucosyltransferase responsible for catalyzing α1,6-fucosylation in mammals, and α1,6-fucosylated glycoproteins are found to participate in various physiopathological processes. However, whether FUT8/α1,6-fucosylation modulates the functions of trophoblastic cells remains elusive. FUT8 in human placenta villi during 6-8 gestational weeks and trophoblastic cells were detected by Western blot and immunofluorescent staining. α1,6-fucosylation in tissues or cells were measured by Lectin LCA (Lens culinaris) fluorescent staining and Lectin blot. FUT8 expression was down-regulated by siRNA transfection in JAR and JEG-3 cells, and cell viability, motility and invasiveness ability were detected by the functional experiments. α1,6-fucosylation of insulin-like growth factor receptor (IGF-1R) was examined by immunoprecipitation, and the amount of phosphorylated IGF-1R was detected in FUT8 down-regulated JAR cells. Human placenta villi and trophoblastic cells expressed FUT8/α1,6-fucosylation. Knockdown FUT8 by siRNA transfection suppressed the proliferation, epithelial-mesenchymal transition, migration and invasion of JAR and JEG-3 cells. Furthermore, we found that FUT8 modified the α1,6-fucosylation of IGF-1R, and regulated IGF-1 dependent activation of IGF-1R, MAPK and PI3K/Akt signaling pathways in JAR cells. Our results implicate a critical role for FUT8 in maintaining the normal functions of trophoblastic cells, suggesting manipulating FUT8 may be an effective approach in pregnancy.