MilliporeSigma
  • SREBP-dependent lipidomic reprogramming as a broad-spectrum antiviral target.
SREBP-dependent lipidomic reprogramming as a broad-spectrum antiviral target.
Nature communications (2019-01-12)
Shuofeng Yuan, Hin Chu, Jasper Fuk-Woo Chan, Zi-Wei Ye, Lei Wen, Bingpeng Yan, Pok-Man Lai, Kah-Meng Tee, Jingjing Huang, Dongdong Chen, Cun Li, Xiaoyu Zhao, Dong Yang, Man Chun Chiu, Cyril Yip, Vincent Kwok-Man Poon, Chris Chung-Sing Chan, Kong-Hung Sze, Jie Zhou, Ivy Hau-Yee Chan, Kin-Hang Kok, Kelvin Kai-Wang To, Richard Yi-Tsun Kao, Johnson Yiu-Nam Lau, Dong-Yan Jin, Stanley Perlman, Kwok-Yung Yuen
ABSTRACT
Viruses are obligate intracellular microbes that exploit the host metabolic machineries to meet their biosynthetic demands, making these host pathways potential therapeutic targets. Here, by exploring a lipid library, we show that AM580, a retinoid derivative and RAR-α agonist, is highly potent in interrupting the life cycle of diverse viruses including Middle East respiratory syndrome coronavirus and influenza A virus. Using click chemistry, the overexpressed sterol regulatory element binding protein (SREBP) is shown to interact with AM580, which accounts for its broad-spectrum antiviral activity. Mechanistic studies pinpoint multiple SREBP proteolytic processes and SREBP-regulated lipid biosynthesis pathways, including the downstream viral protein palmitoylation and double-membrane vesicles formation, that are indispensable for virus replication. Collectively, our study identifies a basic lipogenic transactivation event with broad relevance to human viral infections and represents SREBP as a potential target for the development of broad-spectrum antiviral strategies.