MilliporeSigma
  • A novel thermophilic exochitinase ChiEn3 from Coprinopsis cinerea exhibits a hyperhydrolytic activity toward 85% deacetylated chitosan and a significant application to preparation of chitooligosaccharides from the chitosan.
A novel thermophilic exochitinase ChiEn3 from Coprinopsis cinerea exhibits a hyperhydrolytic activity toward 85% deacetylated chitosan and a significant application to preparation of chitooligosaccharides from the chitosan.
Carbohydrate polymers (2019-01-03)
Jiangsheng Zhou, Rujuan Dai, Yanxin Wang, Maomao Li, Yiting Zhu, Lingling Chen, Liqin Kang, Zhonghua Liu, Yao Yang, Sheng Yuan
ABSTRACT
ChiEn3 from Coprinopsis cinerea was characterized as an exo-acting chitinase with a processivity. ChiEn3 hydrolyzed only soluble chitin and exhibited a hyperhydrolytic activity toward 85% deacetylated chitosan which was 33.6-fold higher than its hydrolytic activity toward glycol chitin. Its maximum hydrolytic activity was observed at 60 °C and retained 66.2% of hydrolytic activity after 60 min incubation at 60 °C. Commercial 85% deacetylated chitosan was degraded by ChiEn3 to a series of COSs with a DP of 2-20 in which COSs with a DP of 3-6 were dominant, whereas, lab-prepared chitosan (FA = 0.65) was degraded by ChiEn3 to COSs with a DP of 2-10 in which the AA dimer was dominant. DPPH-radical-scavenging activity of ChiEn3-digested products of 85% deacetylated chitosan was 3.32-fold higher than that of undigested 85% deacetylated chitosan. Therefore, ChiEn3 shows a valuable advantage for application to the preparation of COSs from commercial 85% deacetylated chitosan.
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
4-Nitrophenyl N,N′-diacetyl-β-D-chitobioside, ≥99% (TLC)
Sigma-Aldrich
Laminarin from Laminaria digitata, polysaccharide substrate for laminarinase
Sigma-Aldrich
Chitin from shrimp shells, practical grade, powder
Sigma-Aldrich
Glycol chitosan, ≥60% (titration), crystalline
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Chitinase Assay Kit, sufficient for 100 multiwell tests