MilliporeSigma
  • Vaccinia virus hijacks EGFR signalling to enhance virus spread through rapid and directed infected cell motility.
Vaccinia virus hijacks EGFR signalling to enhance virus spread through rapid and directed infected cell motility.
Nature microbiology (2018-11-14)
Corina Beerli, Artur Yakimovich, Samuel Kilcher, Glennys V Reynoso, Gotthold Fläschner, Daniel J Müller, Heather D Hickman, Jason Mercer
ABSTRACT
Cell motility is essential for viral dissemination1. Vaccinia virus (VACV), a close relative of smallpox virus, is thought to exploit cell motility as a means to enhance the spread of infection1. A single viral protein, F11L, contributes to this by blocking RhoA signalling to facilitate cell retraction2. However, F11L alone is not sufficient for VACV-induced cell motility, indicating that additional viral factors must be involved. Here, we show that the VACV epidermal growth factor homologue, VGF, promotes infected cell motility and the spread of viral infection. We found that VGF secreted from early infected cells is cleaved by ADAM10, after which it acts largely in a paracrine manner to direct cell motility at the leading edge of infection. Real-time tracking of cells infected in the presence of EGFR, MAPK, FAK and ADAM10 inhibitors or with VGF-deleted and F11-deleted viruses revealed defects in radial velocity and directional migration efficiency, leading to impaired cell-to-cell spread of infection. Furthermore, intravital imaging showed that virus spread and lesion formation are attenuated in the absence of VGF. Our results demonstrate how poxviruses hijack epidermal growth factor receptor-induced cell motility to promote rapid and efficient spread of infection in vitro and in vivo.