- N-Heterocyclic Carbene Complexes of Copper, Nickel, and Cobalt.
N-Heterocyclic Carbene Complexes of Copper, Nickel, and Cobalt.
Chemical reviews (2019-03-08)
Andreas A Danopoulos, Thomas Simler, Pierre Braunstein
PMID30843688
ABSTRACT
The emergence of N-heterocyclic carbenes as ligands across the Periodic Table had an impact on various aspects of the coordination, organometallic, and catalytic chemistry of the 3d metals, including Cu, Ni, and Co, both from the fundamental viewpoint but also in applications, including catalysis, photophysics, bioorganometallic chemistry, materials, etc. In this review, the emergence, development, and state of the art in these three areas are described in detail.