MilliporeSigma
  • Aurora A plays a dual role in migration and survival of human glioblastoma cells according to the CXCL12 concentration.
Aurora A plays a dual role in migration and survival of human glioblastoma cells according to the CXCL12 concentration.
Oncogene (2018-08-08)
Estelle Willems, Matthias Dedobbeleer, Marina Digregorio, Arnaud Lombard, Nicolas Goffart, Paul Noel Lumapat, Jeremy Lambert, Priscilla Van den Ackerveken, Martyna Szpakowska, Andy Chevigné, Felix Scholtes, Bernard Rogister
ABSTRACT
Primary glioblastoma is the most frequent human brain tumor in adults and is generally fatal due to tumor recurrence. We previously demonstrated that glioblastoma-initiating cells invade the subventricular zones and promote their radio-resistance in response to the local release of the CXCL12 chemokine. In this work, we show that the mitotic Aurora A kinase (AurA) is activated through the CXCL12-CXCR4 pathway in an ERK1/2-dependent manner. Moreover, the CXCL12-ERK1/2 signaling induces the expression of Ajuba, the main cofactor of AurA, which allows the auto-phosphorylation of AurA.We show that AurA contributes to glioblastoma cell survival, radio-resistance, self-renewal, and proliferation regardless of the exogenous stimulation with CXCL12. On the other hand, AurA triggers the CXCL12-mediated migration of glioblastoma cells in vitro as well as the invasion of the subventricular zone in xenograft experiments. Moreover, AurA regulates cytoskeletal proteins (i.e., Actin and Vimentin) and favors the pro-migratory activity of the Rho-GTPase CDC42 in response to CXCL12. Altogether, these results show that AurA, a well-known kinase of the mitotic machinery, may play alternative roles in human glioblastoma according to the CXCL12 concentration.