MilliporeSigma
  • Adenosine as an endogenous mediator of hypoxia for induction of vascular endothelial growth factor mRNA in U-937 cells.
Adenosine as an endogenous mediator of hypoxia for induction of vascular endothelial growth factor mRNA in U-937 cells.
Biochemical and biophysical research communications (1994-10-14)
E Hashimoto, K Kage, T Ogita, T Nakaoka, R Matsuoka, Y Kira
ABSTRACT
Adenosine induced by hypoxia exerts various effects via different types of receptors. Recently, hypoxia was shown to be a strong inducer of vascular endothelial growth factor, a secreted endothelial cell specific mitogen. In this report, we studied on effects of adenosine on inducibility of VEGF and possible mediation of hypoxia for its induction in U-937 cells. Hypoxia induced expression of VEGF mRNA with an early peak at 1 hour. 5'-N-ethylcarboxamidoadenosine, an adenosine analog, strongly induced VEGF mRNA, which was inhibited by 3,7-dimethyl-1-propargylxanthine (DMPX), an A2-antagonist. The hypoxic induction was inhibited by adenosine deaminase, 7-(beta-hydroxyethyl)theophylline, a non-selective adenosine receptor antagonist and DMPX. These results suggest that the hypoxic induction of VEGF mRNA is mediated by adenosine via A2-receptor in U-937 cells.