MilliporeSigma
  • Steroids from the Deep-Sea-Derived Fungus Penicilliumgranulatum MCCC 3A00475 Induced Apoptosis via Retinoid X Receptor (RXR)-α Pathway.
Steroids from the Deep-Sea-Derived Fungus Penicilliumgranulatum MCCC 3A00475 Induced Apoptosis via Retinoid X Receptor (RXR)-α Pathway.
Marine drugs (2019-03-22)
Chun-Lan Xie, Duo Zhang, Jin-Mei Xia, Chao-Chao Hu, Ting Lin, Yu-Kun Lin, Guang-Hui Wang, Wen-Jing Tian, Zeng-Peng Li, Xiao-Kun Zhang, Xian-Wen Yang, Hai-Feng Chen
ABSTRACT
Five new ergostanes, penicisteroids D-H (1-5), were isolated from the liquid culture of the deep-sea-derived fungus Penicillium granulatum MCCC 3A00475, along with 27 known compounds. The structures of the new steroids were established mainly on the basis of extensive analysis of 1D and 2D NMR as well as HRESIMS data. Moreover, the absolute configurations of 1 were confirmed unambiguously by the single-crystal X-ray crystallography. Compounds 2 and 4⁻7 showed moderate antiproliferative effects selectively against 12 different cancer cell lines with IC50 values of around 5 μM. Compounds 2 and 6, potent RXRα binders with Kd values of 13.8 and 12.9 μM, respectively, could induce apoptosis by a Retinoid X Receptor (RXR)-α-dependent mechanism by regulating RXRα transcriptional expression and promoting the poly-ADP-ribose polymerase (PARP) cleavage. Moreover, they could inhibit proliferation by cell cycle arrest at the G0/G1 phase.