MilliporeSigma
  • Utilization of agro-industrial waste for production of Transglutaminase from Streptomyces mobaraensis.
Utilization of agro-industrial waste for production of Transglutaminase from Streptomyces mobaraensis.
Bioresource technology (2019-05-12)
Syeda Warisul Fatima, Rameshwar Tiwari, Sunil K Khare
ABSTRACT
This work studied the production of Transglutaminase (TGase) using wheat bran as carbon source. The medium components and culture conditions were optimized by statistical Box-Behnken response surface methodology. The release of active Transglutaminase was enhanced by adding (i) protease to remove pro-region to make inactive enzyme to active form, (ii) Cetyl trimethyl ammonium bromide (CTAB) which facilitated more secretion. Under finally optimized conditions viz. 5 g wheat bran, protease: 39.14 U, magnesium chloride (MgCl2): 0.10 M, CTAB: 0.08% and inoculation size: 2% led to 4-fold (12.949 ± 0.061 IU/g) increased TGase production over that of un-optimized conditions. The application of TGase was shown to be useful in effective casein cross-linking.