MilliporeSigma
  • Histone Deacetylase Inhibitors Dose-Dependently Switch Neutrophil Death from NETosis to Apoptosis.
Histone Deacetylase Inhibitors Dose-Dependently Switch Neutrophil Death from NETosis to Apoptosis.
Biomolecules (2019-05-15)
Hussein J Hamam, Nades Palaniyar
ABSTRACT
Acetylation is an important post translational modification of histone that plays a role in regulation of physiological and pathological process in the body. We have recently shown that the inhibition of histone deacetylases (HDAC) by low concentrations of HDAC inhibitors (HDACis), belinostat (up to 0.25 µM) and panobinostat (up to 0.04 µM) promote histone acetylation (e.g., AcH4) and neutrophil extracellular trap formation (NETosis). Clinical use of belinostat and panobinostat often leads to neutropenia and the in vivo concentrations vary with time and tissue locations. However, the effects of different concentrations of these HDACis on neutrophil death are not fully understood. We considered that increasing concentrations of belinostat and panobinostat could alter the type of neutrophil death. To test this hypothesis, we treated human neutrophils with belinostat and panobinostat in the presence or absence of agonists that promote NOX-dependent NETosis (phorbol myristate acetate or lipopolysaccharide from Escherichia coli 0128) and NOX-independent NETosis (calcium ionophores A23187 or ionomycin from Streptomyces conglobatus). Increasing concentrations of HDACis induced histone acetylation in a dose-dependent manner. ROS analyses showed that increasing concentrations of HDACis, increased the degree of NOX-derived ROS production. Higher levels (>1 µM belinostat and >0.2 µM panobinostat) of AcH4 resulted in a significant inhibition of spontaneous as well as the NOX-dependent and -independent NETosis. By contrast, the degree of neutrophil apoptosis significantly increased, particularly in non-activated cells. Collectively, this study establishes that increasing concentrations of belinostat and panobinostat initially increases NETosis but subsequently reduces NETosis or switches the form of cell death to apoptosis. This new information indicates that belinostat and panobinostat can induce different types of neutrophil death and may induce neutropenia and regulate inflammation at different concentrations.