MilliporeSigma
  • A pilot case-control study on the association between N-acetyl derivatives in serum and first-episode schizophrenia.
A pilot case-control study on the association between N-acetyl derivatives in serum and first-episode schizophrenia.
Psychiatry research (2018-12-24)
Ninghua Huang, Bing Cao, Elisa Brietzke, Caroline Park, Danielle Cha, Zihang Pan, Judy Zhu, Yaqiong Liu, Qing Xie, Jing Zeng, Roger S McIntyre, Jingyu Wang, Lailai Yan
ABSTRACT
N-acetyl group metabolites are a general class of endogenous compounds characterized by a conjugated system consisting of an acetyl group and nitrogen moiety. The aim of our exploratory pilot case-control study is to compare the levels of five N-acetyl derivatives (i.e., N-acetyl-glutamine, N-acetyl-ornithine, N6-acetyl-L-lysine, N-acetyl-putrescine, and N-acetyl-galactosamine) in serum samples between individuals with first-episode schizophrenia and healthy controls (HC). A 1:2 age- and sex- matched pilot case-control study was performed, involving 30 cases of first-episode schizophrenia and 60 HC aged between 18 and 40 years old. The serum samples containing these N-acetyl derivatives from (first-episode patients with schizophrenia and HC were measured using liquid chromatography-tandem mass spectrometry (LC-MS). Results indicated that higher levels of N-acetyl-glutamine and lower levels of N6-acetyl-L-lysine may have a significant association with schizophrenia after adjusting for age, sex and BMI. N-acetyl-putrescine was elevated among subjects with first-episode schizophrenia when compared to HC, suggesting it as a predictor for schizophrenia onset. Further exploration of the mechanisms of N-acetyl group metabolites with respect to schizophrenia is warranted and may be useful for identifying novel disease markers and/or drug target molecules in schizophrenia.