MilliporeSigma
  • Growth Factor Screening in Dystrophic Muscles Reveals PDGFB/PDGFRB-Mediated Migration of Interstitial Stem Cells.
Growth Factor Screening in Dystrophic Muscles Reveals PDGFB/PDGFRB-Mediated Migration of Interstitial Stem Cells.
International journal of molecular sciences (2019-03-08)
Jordi Camps, Hanne Grosemans, Rik Gijsbers, Christa Maes, Maurilio Sampaolesi
ABSTRACT
Progressive muscle degeneration followed by dilated cardiomyopathy is a hallmark of muscular dystrophy. Stem cell therapy is suggested to replace diseased myofibers by healthy myofibers, although so far, we are faced by low efficiencies of migration and engraftment of stem cells. Chemokines are signalling proteins guiding cell migration and have been shown to tightly regulate muscle tissue repair. We sought to determine which chemokines are expressed in dystrophic muscles undergoing tissue remodelling. Therefore, we analysed the expression of chemokines and chemokine receptors in skeletal and cardiac muscles from Sarcoglycan-α null, Sarcoglycan-β null and immunodeficient Sgcβ-null mice. We found that several chemokines are dysregulated in dystrophic muscles. We further show that one of these, platelet-derived growth factor-B, promotes interstitial stem cell migration. This finding provides perspective to an approachable mechanism for improving stem cell homing towards dystrophic muscles.