MilliporeSigma
  • Effects of Free Fatty Acids with Different Chain Lengths and Degrees of Saturability on the Milk Fat Synthesis in Primary Cultured Bovine Mammary Epithelial Cells.
Effects of Free Fatty Acids with Different Chain Lengths and Degrees of Saturability on the Milk Fat Synthesis in Primary Cultured Bovine Mammary Epithelial Cells.
Journal of agricultural and food chemistry (2019-07-16)
Qiongxian Yan, Shaoxun Tang, Chuanshe Zhou, Xuefeng Han, Zhiliang Tan
ABSTRACT
How short-chain fatty acids (FAs) affect cell membrane morphology and milk fat biosynthesis in mammary epithelial cells (MECs) is yet unclear. This study investigated the primary bovine MEC response to different FAs. We observed that the cell surface ultrastructures were influenced by chain length and degree of saturability of FAs. The CD36, FATP1, and FABP3 gene expression was affected independent of the type of FA. FASN, LPIN1, PPARα, and PPARγ transcripts were more sensitive to the short-chain FAs (acetic and β-hydroxybutyric acids). Furthermore, short-chain FAs inclined to regulate FA degradation-, elongation-, and metabolism-associated pathways, while long-chain FAs (stearic and trans-10,cis-12 conjugated linolenic acids) modulated extracellular matrix-receptor interaction-, transcriptional misregulation-, microRNA-, and ribosome biogenesis-related pathways. However, triacylglycerol accumulation in the cytoplasm was not changed by all of the FAs. Overall, FAs with different chain lengths and degrees of saturability could differentially alter primary bovine MEC cell morphology and influence protein profiles involved in milk fat synthesis pathways.