MilliporeSigma
  • Sympathomimetics regulate neuromuscular junction transmission through TRPV1, P/Q- and N-type Ca2+ channels.
Sympathomimetics regulate neuromuscular junction transmission through TRPV1, P/Q- and N-type Ca2+ channels.
Molecular and cellular neurosciences (2019-02-15)
Anna Zaia Carolina Rodrigues, Zhong-Min Wang, María Laura Messi, Osvaldo Delbono
ABSTRACT
Increasing evidence indicates that, first, the sympathetic nervous system interacts extensively with both vasculature and skeletal muscle fibers near neuromuscular junctions (NMJs) and, second, its neurotransmitter, noradrenaline, influences myofiber molecular composition and function and motor innervation. Since sympathomimetic agents have been reported to improve NMJ transmission, we examined whether two in clinical use, salbutamol and clenbuterol, affect the motor axon terminal via extracellular Ca2+ and molecular targets, such as TRPV1 and P/Q- and N-type voltage-activated Ca2+ channels. Electrophysiological recordings in ex-vivo preparations of peroneal nerves and lumbricalis muscles from young adult mice focused on spontaneous miniature end-plate potentials and singly and repetitively evoked end-plate potentials. Adding one dose of salbutamol or clenbuterol to the nerve/muscle preparation or repeatedly administering salbutamol to a mouse for 4 weeks increased spontaneous and evoked synaptic vesicle release but induced a steep decline in EPP amplitude in response to repetitive nerve stimulation. These effects were mediated primarily by ω-agatoxin IVA-sensitive P/Q-type and secondarily by ω-conotoxin GVIA-sensitive N-type Ca2+ channels. Presynaptic arvanil-sensitive TRPV1 channels seem to regulate Ca2+ at the motor neuron terminal at rest, while putative presynaptic β-adrenergic receptors may mediate sympathomimetic and catecholamine effects on presynaptic Ca2+ channels during NMJ activation.