MilliporeSigma
  • Label-free multi-parametric imaging of single cells: dual picosecond optoacoustic microscopy.
Label-free multi-parametric imaging of single cells: dual picosecond optoacoustic microscopy.
Journal of biophotonics (2019-05-31)
Liwang Liu, Laurent Plawinski, Marie-Christine Durrieu, Bertrand Audoin
ABSTRACT
Advances in microscopy with new visualization possibilities often bring dramatic progress to our understanding of the intriguing cellular machinery. Picosecond optoacoustic micro-spectroscopy is an optical technique based on ultrafast pump-probe generation and detection of hypersound on time durations of picoseconds and length scales of nanometers. It is experiencing a renaissance as a versatile imaging tool for cell biology research after a plethora of applications in solid-state physics. In this emerging context, this work reports on a dual-probe architecture to carry out real-time parallel detection of the hypersound propagation inside a cell that is cultured on a metallic substrate, and of the hypersound reflection at the metal/cell adhesion interface. Using this optoacoustic modality, several biophysical properties of the cell can be measured in a noncontact and label-free manner. Its abilities are demonstrated with the multiple imaging of a mitotic macrophage-like cell in a single run experiment.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder