MilliporeSigma
  • The histone chaperone complex FACT promotes proliferative switch of G0 cancer cells.
The histone chaperone complex FACT promotes proliferative switch of G0 cancer cells.
International journal of cancer (2018-12-15)
Ling Bi, Chanlu Xie, Mu Yao, Su Su Thae Hnit, Soma Vignarajan, Yilun Wang, Qian Wang, Zhichao Xi, Hongxi Xu, Zhong Li, Paul de Souza, Andrew Tee, Matthew Wong, Tao Liu, Xiaodong Zhao, Jia Zhou, Ling Xu, Qihan Dong
ABSTRACT
Cancer cell repopulation through cell cycle re-entry by quiescent (G0 ) cell is thought to be an important mechanism behind treatment failure and cancer recurrence. Facilitates Chromatin Transcription (FACT) is involved in DNA repair, replication and transcription by eviction of histones or loosening their contact with DNA. While FACT expression is known to be high in a range of cancers, the biological significance of the aberrant increase is not clear. We found that in prostate and lung cancer cells FACT mRNA and protein levels were low at G0 compared to the proliferating state but replenished upon cell cycle re-entry. Silencing of FACT with Dox-inducible shRNA hindered cell cycle re-entry by G0 cancer cells, which could be rescued by ectopic expression of FACT. An increase in SKP2, c-MYC and PIRH2 and a decrease in p27 protein levels seen upon cell cycle re-entry were prevented or diminished when FACT was silenced. Further, using mVenus-p27K- infected cancer cells to measure p27 degradation capacity, we confirm that inhibition of FACT at release from quiescence suppressed the p27 degradation capacity resulting in an increased mVenus-p27K- signal. In conclusion, FACT plays an important role in promoting the transition from G0 to the proliferative state and can be a potential therapeutic target to prevent prostate and lung cancer from progression and recurrence.