MilliporeSigma
  • Manufacturing the ultimate green banana flour: Impact of drying and extrusion on phenolic profile and starch bioaccessibility.
Manufacturing the ultimate green banana flour: Impact of drying and extrusion on phenolic profile and starch bioaccessibility.
Food chemistry (2019-06-30)
Joana Pico, Kang Xu, Mengmeng Guo, Zulfiqar Mohamedshah, Mario G Ferruzzi, Mario M Martinez
ABSTRACT
Use of banana flours as functional ingredients is growing due to their nutritional benefits derived from phenolics and dietary fiber. However, the effect oven-drying, freeze-drying and extrusion on the phenolic compounds or starch digestibility is not understood. In this work, phenolic acids (gallic acid, caffeic acid), flavan-3-ols (epicatechin, catechin) and flavonols (quercetin-3-O-glucoside and myricetin) were quantified in banana flour processed by different methods. Epicatechin, the most abundant phenolic in all flours (up to 1.93 mg/100 g), was significantly reduced during thermal processing (oven-drying and extrusion). Meanwhile, phenolic acids and flavonols were found to be more thermally stable. Thus, oven-drying and extrusion generally improved the extractability of phenolic acids and flavonols. Freeze-drying resulted in native flours with significantly higher insoluble dietary fiber (up to 43.3%), although the digestible starch fraction was digested more rapidly than the oven-dried counterpart.