MilliporeSigma
  • Doxycycline hyclate: A schistosomicidal agent in vitro with immunomodulatory potential on granulomatous inflammation in vivo.
Doxycycline hyclate: A schistosomicidal agent in vitro with immunomodulatory potential on granulomatous inflammation in vivo.
International immunopharmacology (2019-03-11)
Miriam Viviane Dias, Aline Pereira Castro, Camila Cabral Campos, Thaiany Goulart Souza-Silva, Reggiani Vilela Gonçalves, Raquel Lopes Martins Souza, Marcos José Marques, Rômulo Dias Novaes
ABSTRACT
We investigated the effect in vitro and in vivo of doxycycline hyclate (Dx), a broad-spectrum antibiotic inhibitor of matrix metaloproteinases (MMPs), on adult Schistosoma mansoni worms and granulomatous liver inflammation in infected mice. Adult S. mansoni worms in culture treated with different concentrations of Dx (50-180 μg/mL) were studied for eight days to assess its morphology, eggs production, and mortality. Uninfected mice and those infected with S. mansoni, untreated and treated with praziquantel (Pz; 200 mg/kg) or Dx (50 mg/kg), were evaluated for 60 days. Our results indicated that Dx induced dose-dependent integumentary lesions (bubbles, tubercle collapse, spicule disappearance, peeling, and erosion), reduced mating rate and eggs-laying in adult S. mansoni worms. The effective lethal dose required to kill 50% of worms was 112.0 μg/mL Dx (DL50). In mice, S. mansoni infection induced hepatomegaly, intense IL-4, IL-10, TNF-α and TGF-β production, granulomatous inflammation and hepatic glycogen depletion. The number and size of the granulomas was similar in untreated and Dx-treated animals. Untreated animals showed a predominance of productive granulomas, and intense MMP-2 and MMP-9 activities. Dx-treated mice exhibited a significant increase in IL-4 levels, tissue inflammation, proportion of involutive granulomas, and hepatic collagenogenesis, as well as attenuated MMP-2 and MMP-9 activities. Our findings indicated that Dx is toxic to adult S. mansoni worms in vitro. However, in vitro beneficial effects were not reproduced in vivo, since Dx treatment increased liver granulomatous inflammation and collagenogenesis in S. mansoni-infected mice by a process potentially associated with Dx-mediated hepatic MMP-2 and MMP-9 inhibition.