MilliporeSigma
Asymmetric bromolactonization using amino-thiocarbamate catalyst.
Journal of the American Chemical Society (2010-10-16)
Ling Zhou, Chong Kiat Tan, Xiaojian Jiang, Feng Chen, Ying-Yeung Yeung
ABSTRACT
A novel amino-thiocarbamate-catalyzed bromolactonization of unsaturated carboxylic acids has been developed. The scope of the reaction is evidenced by 22 examples of γ-lactones with up to 99% yield and 93% ee. The protocol was applied in the enantioselective synthesis of the key intermediates of VLA-4 antagonists.
Sigma-Aldrich
Methylboronic acid, 97%