MilliporeSigma
  • Improved hole injection for blue phosphorescent organic light-emitting diodes using solution deposited tin oxide nano-particles decorated ITO anodes.
Improved hole injection for blue phosphorescent organic light-emitting diodes using solution deposited tin oxide nano-particles decorated ITO anodes.
Scientific reports (2019-02-23)
Seung Il Lee, Geum Jae Yun, Jin Wook Kim, Gregory Hanta, Kunyu Liang, Lazar Kojvic, Lok Shu Hui, Ayse Turak, Woo Young Kim
ABSTRACT
Blue phosphorescent organic light-emitting diodes (PHOLEDs) were fabricated with tin oxide (SnOx) nano-particles (NPs) deposited at the ITO anode to improve their electrical and optical performances. SnOx NPs helped ITO to increase the work function enhancing hole injection capability. Charge balance of the device was achieved using p- and n-type mixed host materials in emissive layer and the devices' luminance and maximum external quantum efficiency (EQE) increased about nearly 30%. Tuning the work function using solution processed NPs allows rapid optimization of device efficiency.