MilliporeSigma
  • N4-acetylcytidine is required for sustained NLRP3 inflammasome activation via HMGB1 pathway in microglia.
N4-acetylcytidine is required for sustained NLRP3 inflammasome activation via HMGB1 pathway in microglia.
Cellular signalling (2019-03-12)
JuanJuan Duan, Qifang Zhang, Xiaohong Hu, Deqin Lu, Wenfeng Yu, Hua Bai
ABSTRACT
Persistent inflammasome activation contributes to chronic, low grade inflammation. However, it is unclear how the inflammasome activation is sustained after initiation. Here we reported that N4-acetylcytidine (N4A), a nucleoside metabolite, activated microglia and sustained NLRP3 inflammasome activation by inducing HMGB1 signaling. Released HMGB1 through N4A activated NFκB and induced NLRP3 expression. HMGB1 silencing abolished N4A-stimulated NFκB activation, NLRP3 and persistent HMGB1 expression. In addition, inhibiting NLRP3 expression by RNAi abrogated N4A-mediated HMGB1 expression. Lack of NLRP3 inflammasome adaptor named apoptosis-associated speck-like protein containing a CARD (ASC) abrogated N4A-induced HMGB1 expression, NFκB activation, and NLRP3 expression. Taken together, our results reveal a novel role of N4A in activation of NLRP3 inflamasome via HMGB1 feedback.