MilliporeSigma
  • Prenatal vitamin D and cord blood insulin-like growth factors in Dhaka, Bangladesh.
Prenatal vitamin D and cord blood insulin-like growth factors in Dhaka, Bangladesh.
Endocrine connections (2019-05-10)
Monika Bilic, Huma Qamar, Akpevwe Onoyovwi, Jill Korsiak, Eszter Papp, Abdullah Al Mahmud, Rosanna Weksberg, Alison D Gernand, Jennifer Harrington, Daniel E Roth
ABSTRACT
Fetal growth restriction is linked to adverse health outcomes and is prevalent in low- and middle-income countries; however, determinants of fetal growth are still poorly understood. The objectives were to determine the effect of prenatal vitamin D supplementation on the insulin-like growth factor (IGF) axis at birth, to compare the concentrations of IGF-I in newborns in Bangladesh to a European reference population and to estimate the associations between IGF protein concentrations and birth size. In a randomized controlled trial in Dhaka, Bangladesh, pregnant women enrolled at 17-24 weeks of gestation were assigned to weekly oral vitamin D3 supplementation from enrolment to delivery at doses of 4200 IU/week, 16,800 IU/week, 28,000 IU/week or placebo. In this sub-study, 559 woman-infant pairs were included for analysis and cord blood IGF protein concentrations were quantified at birth. There were no significant effects of vitamin D supplementation on cord blood concentrations of IGF-I (P = 0.398), IGF-II (P = 0.525), binding proteins (BPs) IGFBP-1 (P = 0.170), IGFBP-3 (P = 0.203) or the molar ratio of IGF-I/IGFBP-3 (P = 0.941). In comparison to a European reference population, 6% of girls and 23% of boys had IGF-I concentrations below the 2.5th percentile of the reference population. IGF-I, IGF-II, IGFBP-3 and the IGF-I/IGFBP-3 ratio were positively associated with at least one anthropometric parameter, whereas IGFBP-1 was negatively associated with birth anthropometry. In conclusion, prenatal vitamin D supplementation does not alter or enhance fetal IGF pathways.