MilliporeSigma
  • Picolinic acid as a matrix for laser mass spectrometry of nucleic acids and proteins.
Picolinic acid as a matrix for laser mass spectrometry of nucleic acids and proteins.
Rapid communications in mass spectrometry : RCM (1994-09-01)
K Tang, N I Taranenko, S L Allman, C H Chen, L Y Cháng, K B Jacobson
ABSTRACT
We found that picolinic acid is a very good matrix for matrix-assisted laser desorption/ionization (MALDI) time-of-flight mass spectrometry of oligonucleotides, proteins and tRNA. Among the oligonucleotides, picolinic acid was shown to be effective for homo-oligonucleotides, d(G)40 and d(C)60, and for mixed-base oligonucleotides up to 190 bases. In the case of the single-stranded oligonucleotides, and of double-stranded ones as well, only parent ions corresponding to single-stranded DNA were observed. The efficiency for MALDI of oligonucleotides using the picolinic acid matrix was superior to that using 3-hydroxypicolinic acid. MALDI of transfer RNA phenylalanine-specific (tRNA(Phe)), a 76-base ribonucleic acid, was detected with a signal-to-noise ratio of > 10. By comparison with 3-hydroxypicolinic acid the results with picolinic acid are notably superior for all oligonucleotides.
Sigma-Aldrich
3-Hydroxypyridine-2-carboxylic acid, 98%
Supelco
3-Hydroxypicolinic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)